Learn More
Biotium CF594 TUNEL Assay Apoptosis Detection Kit
Supplier:  Biotium 30064
Encompass_Preferred
In the TUNEL assay, TdT enzyme catalyzes the addition of labeled dUTP to the 3'- ends of the internucleosomal DNA strand breaks that are a hallmark of apoptosis. The CF dye TUNEL Assay Apoptosis Detection Kit employs dUTP conjugated to Biotium's exceptionally bright and photostable CF fluorescent dyes, for single step fluorescent TUNEL labeling of fixed cells or tissues for analysis by fluorescence microscopy or flow cytometry. This kit contains CF594 which has an excitation/emission of 593/614 nm. The kit has reagents sufficient for 50 reactions.
Catalog No. 50-196-4491