Learn More
Biotium CF Dye Dextran 70,000 MW, Anionic and Fixable, 1mg
Supplier:  Biotium 80129
Encompass_Preferred
This dextran is 70,000 MW, and contains a formaldehyde-fixable free-amine group; available with a selection of CF Dyes. CF Dye labeled dextrans could be used as a fluorescent fluid-phase markers to study cell permeability, endocytosis, or mechanisms of biomolecular delivery. The dextran is 70,000 MW, and contains a formaldehyde-fixable free-amine group.
Catalog No. 50-196-4796