Learn More
Selleck Chemical LLC CFI-400945 5mg 1338806-73-7
SDP
Supplier:  Selleck Chemical LLC S75525MG
Encompass_Preferred
CFI-400945 is an orally active, potent and selective polo-like kinase 4(PLK4) inhibitor with Ki value of 0.26 nM. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4937