Learn More
Cytek Biosciences Inc cFluor B515 aHu HLA-DR Anti
Supplier: Cytek Biosciences Inc R720101
Product Name: cFluor B515 Anti-Human HLA-DR (L243)
Product Class: Antibody
Type: Primary
Product Name: cFluor B515 Anti-Human HLA-DR (L243)
Cytek Catalog Number: R7-20101
Marker: HLA-DR
Target Species: Human
Reported Cross Reactivity: African Green Monkey|Baboon|Chimpanzee|Cynomolgus Monkey|Dog|Common Marmoset|Rhesus Macaque|Squirrel Monkey
Clonality: Monoclonal
Clone: L243
Host Species: Mouse
Isotype: IgG2a, k
Conjugation: cFluor B515
Size: 100 Tests
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.