Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc cFluor R720 aHu Mu CD44 Anti

Supplier:  Cytek Biosciences Inc R720562

Encompass_Preferred

Product Name: cFluor R720 Anti-Human/Mouse CD44 (IM7)
Product Class: Antibody
Type: Primary
Product Name: cFluor R720 Anti-Human/Mouse CD44 (IM7)
Cytek Catalog Number: R7-20562
Marker: CD44
Target Species: Human|Mouse
Reported Cross Reactivity: Baboon|Chimpanzee|Cynomolgus Monkey|Rhesus Macaque|Equine|Bovine|Pig|Dog|Feline
Clonality: Monoclonal
Clone: IM7
Host Species: Rat
Isotype: IgG2b, k
Conjugation: cFluor R720
Size: 25 Tests

Catalog No. 50-000-87173


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.