Learn More
Cytek Biosciences Inc cFluor B548 aMu Ly-6G Anti
Supplier: Cytek Biosciences Inc R720543
Product Name: cFluor B548 Anti-Mouse Ly-6G (1A8)
Product Class: Antibody
Type: Primary
Product Name: cFluor B548 Anti-Mouse Ly-6G (1A8)
Cytek Catalog Number: R7-20543
Marker: Ly-6G
Target Species: Mouse
Clonality: Monoclonal
Clone: 1A8
Host Species: Rat
Isotype: IgG2a, k
Conjugation: cFluor B548
Size: 100 Tests
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.