Learn More
Cytek Biosciences Inc cFluor BYG575 aMu CD64 Anti
Supplier: Cytek Biosciences Inc R720569
Product Name: cFluor BYG575 Anti-Mouse CD64 (X54-5/7.1)
Product Class: Antibody
Type: Primary
Product Name: cFluor BYG575 Anti-Mouse CD64 (X54-5/7.1)
Cytek Catalog Number: R7-20569
Marker: CD64
Target Species: Mouse
Clonality: Monoclonal
Clone: X54-5/7.1
Host Species: Mouse
Isotype: IgG1, k
Conjugation: cFluor BYG575
Size: 100 Tests
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.