Learn More
Cytek Biosciences Inc cFluor BYG710 aMu CD49b Anti
Supplier: Cytek Biosciences Inc R720578
Product Name: cFluor BYG710 Anti-Mouse CD49b (HMa2)
Product Class: Antibody
Type: Primary
Product Name: cFluor BYG710 Anti-Mouse CD49b (HMa2)
Cytek Catalog Number: R7-20578
Marker: CD49b
Target Species: Mouse
Clonality: Monoclonal
Clone: HMa2
Host Species: Armenian Hamster
Isotype: IgG
Conjugation: cFluor BYG710
Size: 25 Tests
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.