Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc cFluor™ BYG750 Anti-Human CD4, 20 μg / mL, 100 tests, 5 μL / test, Clone SK3, Mouse IgG1, κ, Flow cytometry, Human

Supplier:  Cytek Biosciences Inc R720159

Encompass_Preferred

The cFluor™ BYG750 Anti-Human CD4 antibody from Cytek Biosciences is tailored for effective flow cytometric analysis of human CD4. It is conjugated to the cFluor™ BYG750 fluorophore, making it compatible with full spectrum flow cytometry platforms. This monoclonal antibody specifically targets human CD4, essential for immune response and HIV research. Each lot undergoes rigorous quality control testing, ensuring reliability in research applications.

  • Designed for flow cytometry
  • Targets human CD4 with high specificity
  • Ensures accurate detection of CD4 in research
  • Conjugated with cFluor™ BYG750 for enhanced performance
  • Quality control tested via flow cytometry
  • Phosphate-buffered formulation for stability
  • Not intended for diagnostic use
  • Supplied in a ready-to-use format
  • Storage conditions: 2-8°C, protected from light

Catalog No. NC3371304


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.