Learn More
Cytek Biosciences Inc cFluor V435 aMu CD25 Anti
Supplier: Cytek Biosciences Inc R721033
Product Name: cFluor V435 Anti-Mouse CD25 (PC61.5.3)
Product Class: Antibody
Type: Primary
Product Name: cFluor V435 Anti-Mouse CD25 (PC61.5.3)
Cytek Catalog Number: R7-21033
Marker: CD25
Target Species: Mouse
Clonality: Monoclonal
Clone: PC61.5.3
Host Species: Rat
Isotype: IgG1, l
Conjugation: cFluor V435
Size: 100 Tests
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.