Learn More
Cytek Biosciences Inc cFluor V475 aHu CD3 Anti
Supplier: Cytek Biosciences Inc R721043
Product Name: cFluor V475 Anti-Human CD3 (UCHT1)
Product Class: Antibody
Type: Primary
Product Name: cFluor V475 Anti-Human CD3 (UCHT1)
Cytek Catalog Number: R7-21043
Marker: CD3
Target Species: Human
Reported Cross Reactivity: Chimpanzee
Clonality: Monoclonal
Clone: UCHT1
Host Species: Mouse
Isotype: IgG1, k
Conjugation: cFluor V475
Size: 100 Tests
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.