Learn More
Cytek Biosciences Inc cFluor V570 aHu CD69 Anti
Supplier: Cytek Biosciences Inc R721109
Product Name: cFluor V570 Anti-Human CD69 (FN50)
Product Class: Antibody
Type: Primary
Product Name: cFluor V570 Anti-Human CD69 (FN50)
Cytek Catalog Number: R7-21109
Marker: CD69
Target Species: Human
Clonality: Monoclonal
Clone: FN50
Host Species: Mouse
Isotype: IgG1, k
Conjugation: cFluor V570
Size: 100 Tests
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.