Learn More
Cytek Biosciences Inc cFluor V605 aHu CD14 Anti
Supplier: Cytek Biosciences Inc R721123
Product Name: cFluor V605 Anti-Human CD14 (TÜK4)
Product Class: Antibody
Type: Primary
Product Name: cFluor V605 Anti-Human CD14 (TÜK4)
Cytek Catalog Number: R7-21123
Marker: CD14
Target Species: Human
Reported Cross Reactivity: Non-Human Primate|Rhesus Macaque|Cynomolgus Monkey|Pigtail Monkey|Cotton-Topped Tamarin|Pig|Bovine|Ovine|Goat|Dog|Mink|Rabbit
Clonality: Monoclonal
Clone: TÜK4
Host Species: Mouse
Isotype: IgG2a
Conjugation: cFluor V605
Size: 100 Tests
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.