Learn More
TARGETMOL CHEMICALS INC CFT7455 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T600245MG
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. CFT7455 (Cemsidomide) is an anti-cancer agent showing high affinity to the cereblon E3 ligase with a Kd of 0.9 nM. CFT7455 is an orally active degrader of zinc finger transcription factors Ikaros (IKZF1), and Aiolos (IKZF3). Purity 97.93%
Catalog No. 50-288-0031