Learn More
TARGETMOL CHEMICALS INC CG 428 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T412245MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CG 428 is a potent tropomyosin receptor Kinase (TRK) Degrader (uSMITETM) with a DC50 of 0.36 nM. CG 428 comprises an analog of the pan-TRK inhibitor GNF-8625 joined by a linker to the cereblon E3 ligase ligand pomalidomide. CG 428 shows selectivity for TRKA over TRKC and TRKB with Kds of 1nM 4.2 nM and 28 nM. CG 428 inhibits the growth of KM12 colon cancer cells with an IC50 of 2.9 nM. purity: 98%
Catalog No. 50-257-9949