Learn More
Medchemexpress LLC CG347B | 1598426-03-9 | 98.1% | C16H17N3O2 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13589010MG
Encompass_Preferred
CG347B is a selective small-molecule inhibitor of histone deacetylase 6 (HDAC6) intended for research use. It is applied in studies of oncology, immunology, and neurology and can be used as an intermediate in the synthesis of metalloenzyme inhibitors. The compound is supplied with characterization data to support experimental reproducibility.
  • Selective inhibition of HDAC6 enabling targeted biological studies.
  • Small-molecule structure suitable for cellular and biochemical assays.
  • High purity for reproducible experimental results.
  • Documented solubility in DMSO for assay preparation.
  • Applicable to oncology, immunology, and neurology research models.
  • Suitable as a synthetic intermediate for metalloenzyme inhibitor development.
Catalog No. 50-002-61332