Learn More
Cayman Chemical cGAS TR-FRET asy Buffer4X 1ea
SDP
Supplier:  Cayman Chemical 4005191 EA
Encompass_Preferred
cGAS TR-FRET Assay Buffer (4X) has been tested and formulated to work exclusively with Cayman’s cGAS TR-FRET Inhibitor Screening Assay Kit (Item No. 702120).
Catalog No. 50-294-3191