Learn More
Medchemexpress LLC CGI-1746 Btk Inhibitor | 910232-84-7 | 99.39% | 579.69 | 5 MG

Supplier: Medchemexpress LLC 5MGHY11999
CGI-1746 is a potent and highly selective inhibitor of Btk with an IC50 of 1.9 nM. This compound is for research use only and not sold to patients.
- Potent and highly selective Btk inhibitor
- IC50 of 1.9 nM for Btk
- Purity of 99.39%
- Molecular weight is 579.69
- Appears as a white to light yellow solid
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.