Learn More
Medchemexpress LLC cGMP-HTL | 99.7% | 844.27 g/mol | C31H51ClN7O14PS | 5 MG

Supplier: Medchemexpress LLC HY1338695MG
cGMP-HTL (cGMP-HaloTag-ligand) is a cGMP-containing HaloTag ligand developed as an autophagy inducer. It promotes K63-linked ubiquitination of mitochondria and is supplied as a high-purity solid with recommended solvent solubility and storage conditions for research use.
- Induces autophagy by increasing K63-linked ubiquitination of mitochondria.
- Contains an HT-ligand, a linker, and a Cys-S-cGMP autophagy tag.
- High purity (99.7%) suitable for biochemical and cell-based studies.
- Soluble in DMSO at 100 mg/mL; ultrasonic assistance recommended.
- Store sealed, away from moisture and light; in solution store at -80°C up to 6 months or -20°C up to 1 month.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.