Learn More
American Radiolabeled Chemicals Inc CGP 12177 3H 250UC
SDP
Supplier:  American Radiolabeled Chemicals Inc ART 1752250UC
Encompass
(-)-CGP 12177-5,7-3H 250uCi in ethanol, 1 mCi/ ml, S.A. 30-60 Ci/mmol, M.W. 279.80, Shipped in Dry Ice
Catalog No. 50-901-102