Learn More
Medchemexpress LLC Cgp 12177 hydrochloride | 64208-32-8 | MFCD00069230 | 99.7% | 315.80 | C14H22ClN3O3 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY101393A10MG
Encompass_Preferred
CGP 12177 hydrochloride is a research-grade β-adrenergic receptor ligand commonly used in cardiovascular and metabolic disease studies to probe adrenergic receptor function. It is provided as the hydrochloride salt with high reported purity for laboratory research.
  • Acts as a β1/β2 antagonist and partial β3 agonist.
  • Suitable for in vitro and in vivo pharmacology studies.
  • High reported purity for consistent experimental results.
  • Available in small milligram packages for precise dosing in experiments.
Catalog No. 50-002-12464