Learn More
TARGETMOL CHEMICALS INC CGP 20376 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T308195MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg and bulk. Please contact Fisher for quotes. CGP 20376 is a benzothiazole compound an anthelmintic with dual dose-dependent modulation of in vitro effects on the respiratory burst of eosinophils and antifibrillatory properties. purity: 91%
Catalog No. 50-258-0388