Learn More
Enzo Life Sciences CGP-37157 (5mg). CAS: 75450-34-9
SDP
Supplier:  Enzo Life Sciences BMLCM1190005
Encompass_Preferred
Potent, selective inhibitor of mitochondrial Na+Ca2+ exchange. IC50=0.4µM. Blocks mitochondrial permeability transition. Purity: ≥98% (TLC). Solubility: Soluble in DMSO. Long Term Storage: Ambient.
Catalog No. 50-201-0424