Learn More
Medchemexpress LLC Cgp-42112 (Cgp-42112A) | 127060-75-7 | MFCD00133611 | 99.8% | 1052.19 g·mol-1 | C52H69N13O11 | 1MG

Supplier: Medchemexpress LLC HY124051MG
CGP-42112 (CGP-42112A) is a peptide-like research compound that acts as a potent angiotensin II subtype 2 (AT2) receptor agonist. It is supplied for research use only and is available as solid material and as a DMSO solution. The product has a defined purity, molecular weight, solubility profile, and recommended storage conditions, making it suitable for in vitro assays and for formulation in common in vivo vehicles.
- Potent angiotensin II subtype 2 receptor agonist.
- High purity (99.84%).
- Molecular weight 1052.19 g·mol-1.
- Soluble in DMSO (100 mg/mL); compatible with common in vivo vehicles.
- Supplied as solid and as DMSO solution in multiple pack sizes.
- Powder stable at -20 °C for long-term storage; solutions stable at -80 °C.
- For research use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.