Learn More
American Radiolabeled Chemicals Inc CGP 42112A-125I 5UC
SDP
Supplier:  American Radiolabeled Chemicals Inc ARI 01885UC
Encompass
CGP 42112A-125I 5uCi in buffered solution, 0.03 mCi/ml, S.A. 2200 Ci/mmol, M.W. 1176
Catalog No. 50-901-064
This product is regulated by the federal government, and may not be ordered online.