Learn More
Apexbio Technology LLC CGP 52432 139667-74-6 10mg
SDP
Supplier:  Apexbio Technology LLC B665510
Encompass_Preferred
CGP 52432 (CAS 139667-74-6) is a selective antagonist of the GABAB receptor exhibiting an IC50 of 85 nM By inhibiting GABAB receptor-mediated signaling pathways CGP 52432 modulates neurotransmitter release and alters neuronal activity This compound is widely utilized in neuroscience research to investigate GABAergic mechanisms underlying physiological and pathological processes and to support the development of therapeutic strategies targeting GABAB receptor function
Catalog No. 50-000-88413