Learn More
Medchemexpress LLC Cgp35348 | 123690-79-9 | MFCD11045877; MFCD00882736 | >98.0% | 225.22 g/mol | C8H20NO4P | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10353010MM 1ML
Encompass_Preferred
CGP35348 (CAS 123690-79-9) is a selective GABAB receptor antagonist commonly used as a research standard in neuroscience and pharmacology studies. It is supplied by chemical vendors as an analytical standard (solid) and, in some supplier SKUs, as prepared solutions for experimental use.
  • Selective GABAB receptor antagonist used in receptor pharmacology studies.
  • Supplied as an analytical standard suitable for assay development.
  • Available in small solid quantities; solution formats may be offered by some suppliers.
  • Used in in vitro and in vivo studies to probe GABAB-mediated signaling.
Catalog No. 50-002-48193