Learn More
Medchemexpress LLC Cgp71683 hydrochloride | 192322-50-2 | MFCD07783999 | ≥98.0% | 512.07 | C26H30ClN5O2S | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10772325MG
Encompass_Preferred
CGP71683 hydrochloride is the hydrochloride salt of a selective, non-peptide antagonist of the neuropeptide Y (NPY) Y5 receptor, supplied for research use in receptor pharmacology and neuroscience studies. It is typically used in cellular assays and biochemical experiments to investigate Y5-mediated signaling pathways.
  • Hydrochloride salt form improves solubility for assay preparation.
  • High purity (>98% by HPLC) suitable for research applications.
  • Potent Y5 receptor antagonism useful for pharmacological profiling.
  • Offered in small lab-scale pack sizes for screening and method development.
  • Characterized by CAS number 192322-50-2 and molecular formula C26H30ClN5O2S.
Catalog No. 50-002-40794