Learn More
Selleck Chemical LLC CGP71683 hydrochloride S0124-25MG
SDP
Supplier:  Selleck Chemical LLC S012425MG
Encompass_Preferred
CGP71683 hydrochloride (CGP71683A) is a competitive and sellective neuropeptide Y receptor type 5 (Y5 receptor) antagonist with Ki of 1 3 nM
Catalog No. 50-000-39807