Learn More
Medchemexpress LLC CGP77675 | 234772-64-6 | 99.5% | 443.54 g·mol⁻¹ | C26H29N5O2 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW06283550MG
Encompass_Preferred
CGP77675 is a small-molecule inhibitor of Src family kinases used in biochemical and cell-based research. It inhibits phosphorylation of peptide substrates and autophosphorylation of c-Src, and shows low-nanomolar potency against Src family members.
  • Potent Src family kinase inhibitor with low-nanomolar activity.
  • Useful for studying signal transduction and cell biology pathways.
  • Reported activity against Src, Lck, and c-Yes.
  • Provided as a solid with high purity for research use.
  • Commonly used in assays investigating bone resorption and epithelial tight junctions.
Catalog No. 50-002-10552