Learn More
Medchemexpress LLC α-CGRP (mouse, rat) | 83651-90-5 | 99.8% | 3806.25 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP02035MG
Encompass_Preferred
α-CGRP (mouse, rat) is a neuropeptide (calcitonin gene-related peptide (CGRP)) mainly expressed in the neuromuscular junction, acting as a potent vasodilator. It can lead to a fall in blood pressure, an increase in heart rate by peripheral administration, and relaxation of colonic smooth muscle. This peptide has potential in cardiovascular, pro-inflammatory, migraine, and metabolic studies.
  • Regulates innate lymphoid cell response and insulin secretion.
  • Decreases mean arterial blood pressure and increases heart rate in vivo.
  • Plays a role in regulating Kainic acid (KA) induced pyramid-cell death in the hippocampal CA3 region.
Catalog No. 50-004-25644