Learn More
Medchemexpress LLC HY-P0203A 1mg Medchemexpress, α-CGRP, rat (TFA) CAS: Purity:>98%
Supplier: Medchemexpress LLC HYP0203A1MG
Medchemexpress, HY-P0203A 1mg α-CGRP, rat (TFA) CAS: α-CGRP, rat TFA, a neuropeptide (calcitonin gene-related peptide (CGRP)), is a potent vasodilator, with the potential in cardiovascular, pro-inflammatory and metabolic studies. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.