Learn More
Medchemexpress LLC CH-223191 | 301326-22-7 | 99.8% | 200 MG

Supplier: Medchemexpress LLC HY12684200MG
CH-223191 is a potent and specific antagonist of aryl hydrocarbon receptor (AhR). It inhibits TCDD-mediated nuclear translocation and DNA binding of AhR, and also inhibits TCDD-induced luciferase activity with an IC50 of 0.03 μM.
- Potent and specific antagonist of aryl hydrocarbon receptor (AhR)
- Inhibits TCDD-mediated nuclear translocation and DNA binding of AhR
- Inhibits TCDD-induced luciferase activity with an IC50 of 0.03 μM
- Inhibits TCDD-caused cytochrome P450 1A1 mRNA expression
- Suppresses cytochrome P450 1A1 expression in liver
- Reduces activity of AST and ALT
- Molecular weight: 333.39
- Molecular formula: C19H19N5O
- Appearance: solid, yellow to orange
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.