Learn More
Cayman Chemical CH 275acetate 10mg
SDP
Supplier:  Cayman Chemical 4105010 MG
Encompass_Preferred
A cyclic peptide agonist of SST1 (IC50 = 30.9 nM for the human receptor); selective for SST1 over SST2, SST3, SST4, and SST5 (IC50s = >10,000, 345, >1,000, and >10,000 nM, respectively, for the human receptors); inhibits basal growth hormone secretion from primary mouse pituitary cells at 500 nM; inhibits glucose-induced increases in electrical activity without inducing membrane hyperpolarization in a whole-cell patch-clamp assay using MIN6 pancreatic β-cells at 100 nM
Catalog No. 50-294-3324