Learn More
Medchemexpress LLC CH1055 1MG | 1622250-38-7 | 94.2% | 969.09 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYD14641MG
Encompass_Preferred
CH1055 is a near-infrared II (NIR-II) fluorescent dye, suitable for use as a fluorescent probe in in vivo imaging applications. It can be conjugated with antibodies or ligands for specific imaging, such as tumor imaging.
  • Much smaller than the renal excretion threshold
  • Easily excreted from the body through the kidneys
  • Reduces toxicity risk from long-term accumulation
  • Excitation wavelength of 808 nM
  • Emission wavelength of 1055 nM
Catalog No. 50-004-31432