Learn More
Medchemexpress LLC CH3OCO-D-CHA-Gly-Arg-pNA acetate | 80895-10-9 | C27H42N8O9 | 1 ML

Supplier: Medchemexpress LLC HY15382910MM/1ML
CH3OCO-D-CHA-Gly-Arg-pNA acetate is a chromogenic substrate for factor Xa. It has a molecular weight of 622.67 and a purity of 99.24%.
- Chromogenic substrate for factor Xa
- Target: Factor Xa
- Pathway: Metabolic enzyme/protease
- Solid storage: 4°C, sealed, away from moisture
- In solvent storage: -80°C for 6 months; -20°C for 1 month (sealed, away from moisture)
- Solubility in DMSO: 250 mg/mL (401.50 mM)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.