Learn More
Medchemexpress LLC CH3OCO-D-CHA-Gly-Arg-pNA acetate | 80895-10-9 | C27H42N8O9 | 25 MG

Supplier: Medchemexpress LLC 25MGHY153829
CH3OCO-D-CHA-Gly-Arg-pNA acetate is a chromogenic substrate for factor Xa, appearing as a white to off-white solid with high purity. It is intended for research use only and has a molecular weight of 622.67.
- Chromogenic substrate for factor Xa
- White to off-white solid appearance
- Purity of 99.24%
- Molecular weight of 622.67
- Storage at 4°C, sealed, away from moisture
- In solvent storage: -80°C for 6 months; -20°C for 1 month (sealed, away from moisture)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.