Learn More
Selleck Chemical LLC Chaetocin 1mg 28097-03-2
SDP
Supplier:  Selleck Chemical LLC S80681MG
Encompass_Preferred
Chaetocin, a natural product from Chaetomium species, is a histone methyltransferase inhibitor with IC50 of 0.8 ?M, 2.5 ?M and 3 ?M for dSU(VAR)3-9, mouse G9a and Neurospora crassa DIM5, respectively. Chaetocin is an anticancer agent and inhibitor of thioredoxin reductase (TrxR). *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5625