Learn More
Cayman Chemical Chaetomela sp WF-3681 100ug
SDP
Supplier:  Cayman Chemical 33523100 UG
Encompass_Preferred
A fungal metabolite; inhibits aldose reductase (IC50 = 0.25 µM)
Catalog No. 50-295-0682