Learn More
Cayman Chemical ChaetomIum AureonItol 100ug
SDP
Supplier:  Cayman Chemical 32925100 UG
Encompass_Preferred
A fungal metabolite; inhibits replication of the influenza A subtype H3N2 at an MOI of 0.01 (EC50 = 30 nM); inhibits replication of laboratory-adapted and clinical isolates of H3N2 (EC50s = 100 and 312 nM, respectively), as well as clinical isolates of H1N1 (EC50 = 417 nM)
Catalog No. 50-295-0548