Learn More
MSC CAT Chainsaw Bar & Chain, 18 in, 3/8 LP, .043 in, 62 Drive Links
Supplier:  MSC 64944358
Encompass
This chainsaw bar and chain serves as a replacement for 18 in chainsaws utilizing a .043 3/8 LP 62 drive link chain. It is engineered for situations of extreme wear or damage and incorporates a small diameter nose radius to enhance safety by mitigating kickback potential.
  • Replacement chainsaw bar and chain for 18 in chainsaws
  • Narrow gauge .043 3/8 LP cutting chain with 62 drive links
  • Designed for enhanced cutting speed and reduced power consumption
  • Small diameter nose radius to increase safety by reducing kickback potential
  • Includes an 18 in .043 gauge chainsaw bar with a 3/8 LP nose sprocket
Catalog No. 50-283-6828