Learn More
FUJIFILM BIOSCIENCES INC CHANG Medium BMC, 100 mL
Supplier:  FUJIFILM BIOSCIENCES INC 91004100ML
Encompass
CHANG Medium BMC was developed for the primary culture of Clinical Human Bone Marrow cultures for use in karyotyping and other genetic testing using existing protocols based on RPMI Medium 1640, 10% GCT-CM and gentamicin.
Catalog No. NC1151202
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.