Learn More
Medchemexpress LLC Chaps | 75621-03-3 | C32H58N2O7S | 500 MG
SDP
Supplier:  Medchemexpress LLC HY15435500MG
Encompass_Preferred
Chaps is a cholic acid-derived, sulfobetaine-type zwitterionic detergent and micelle-forming agent. It exhibits properties of weak cationic or nonionic surfactants in various solution systems, undergoing micellization and forming small, loose hydrophilic aggregates. This substance is commonly employed in research concerning the separation and purification of membrane proteins.
  • Stabilizes mononucleosomes under differing ionic strengths and reduces nucleosome sequence specificity.
  • Facilitates the sliding of histone cores along DNA.
  • Solubilizes Tamm-Horsfall protein, reducing interference with urinary exosome isolation.
  • Maintains vesicle structure and the activity of related proteins.
  • Used for recovering native folded fusion proteins and enhancing the binding capacity of GST fusion proteins.
  • Restores GST enzyme activity.
Catalog No. 50-003-76064