Learn More
Research Products International Corp CHAPSO, 1 G

Supplier: Research Products International Corp C925001.0
Molecular Formula: C32H58N2O8S
Molecular Weight: 630.88
CAS Number: 82473-24-3
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.