Learn More
Cobex Recorders Inc Chart Recorder (C901), 6-inch, Single Pen, Fully Cased
Supplier: Cobex Recorders Inc C6XXXCG0011Y0
6 Inch, Single Pen, Fully Cased. These recorders are highly versatile single process recording instruments. The variety of input signal types, coupled with ease of use, make these recorders ideal monitoring devices. Inherent reliability and precision assure accurate results. These recorders are well suited for the measurement of temperature, humidity, pressure, force, level, time of operation, carbon dioxide, PH and multiple voltage / current signals.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.