Learn More
Bon Opus Biosciences Recombinant Human CHCHD2 Protein(C-His) - Source: E. coli
SDP
Supplier:  Bon Opus Biosciences BP261550UG
Encompass_Preferred
Recombinant Human CHCHD2 Protein is produced by E.coli expression system and the target gene encoding P2-A151 is expressed with a His tag at the C-terminus. Accession#Q9Y6H1. Formulation: Supplied as a 0.2 um filtered solution of 500mM PBS, 300mM NaCl, 10%Glycerol, pH7.4
Catalog No. NC2733752