Learn More
Medchemexpress LLC Chdi-390576 | 1629729-98-1 | 98.1% | 10 MG
SDP
Supplier:  Medchemexpress LLC HY11993910MG
Encompass_Preferred
A small-molecule, cell-permeable, CNS-penetrant class IIa histone deacetylase (HDAC) inhibitor for research use. It is suitable for biochemical and cellular assays to study HDAC biology and related pathways. Available as a solid and in ready-to-use DMSO solutions with recommended cold storage.
  • Potent inhibition of class IIa HDAC isoforms (reported IC50s: HDAC4 54 nM, HDAC5 60 nM, HDAC7 31 nM, HDAC9 50 nM).
  • High selectivity over class I and other HDAC isoforms (>500-fold vs class I; ~150-fold vs HDAC8 and HDAC6).
  • High purity supply (approximately 98.1%).
  • Supplied as powder and as 10 mM solutions in DMSO; recommended storage: powder -20°C, in solvent -80°C (short term at -20°C).
  • Intended for research use only; not for human or diagnostic applications.
Catalog No. 50-002-34652