Learn More
Selleck Chemical LLC Chebulagic acid-E0226-1MG
SDP
Supplier:  Selleck Chemical LLC E02261MG
Encompass_Preferred
Chebulagic acid isolated form Terminalia chebula Retz is a reversible and non-competitive inhibitor of maltase with a Ki value of 6 6 M Chebulagic acid shows potent anti-inflammatory effects in LPS-stimulated RAW 264 7 cells Chebulagic acid also shows potent COX-LOX dual inhibition activity with IC50 values of 15 M 0 92 M and 2 1 M for COX-1 COX-2 and 5-LOX respectively
Catalog No. 50-313-7941