Learn More
Electron Microscopy Sciences Check Temp 4 Complete with Certificate
SDP
Supplier:  Electron Microscopy Sciences 720862C
Encompass_Preferred
Checktemp 4 thermometer is fast responding, fold-away probe is made of high quality, stainless steel. It enables angled measurements and can penetrate semi-frozen and semi-solid samples. The LCD on the side of the handle is easy to see and read.
Unfold the stainless steel probe, the Checktemp 4 automatically turns ON and immediately performs a calibration test and also lets you know if your battery level is low or if your meter requires recalibration. These unique features provide the security of knowing you have accurate measurements. It also features auto shut-off. Calibrated against NIST standards.
Catalog No. 50-189-9744