Learn More
Waters Corp Check Valve Inlet or Outlet Housing, HPLC Pump Component, for Gradient Mixers
Supplier:  Waters Corp WAT055845
Encompass_Preferred
This check valve inlet or outlet housing is a component designed for use with HPLC pumps, specifically within the fluidic connections of gradient mixer configurations. It plays a role in the setup of dual gradient mixers, such as the GM150, for efficient operation.
  • Functions in fluidic connections for HPLC pumps.
  • Associated with the 1525μ Binary HPLC Pump.
  • Integral to the configuration of gradient mixers.
  • Supports setup with dual GM150 Gradient Mixers.
Catalog No. 50-793-657